Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrobenzoyl chloride, 98%

Catalog No. AC150432500
Change view
Click to view available options
Quantity:
5 g
100 g
250 g
500 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC150432500 250 g Glass bottle
AC150430050 5 g Glass bottle
AC150431000 100 g Glass bottle
AC150435000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC150432500 Supplier Thermo Scientific Chemicals Supplier No. 150432500
Only null left
Add to Cart

CAS: 122-04-3 | C7H4ClNO3 | 185.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-04-3
Molecular Formula C7H4ClNO3
Molecular Weight (g/mol) 185.56
MDL Number MFCD00007345
InChI Key SKDHHIUENRGTHK-UHFFFAOYSA-N
Synonym p-nitrobenzoyl chloride, benzoyl chloride, 4-nitro, 4-nitrobenzoic acid chloride, p-nitrobenzoic acid chloride, benzoyl chloride, p-nitro, 4-nitro-benzoyl chloride, unii-x3h8pw2gc4, ccris 3136, 4-nitrobenzoylchloride, 4-nitro benzoyl chloride
PubChem CID 8502
IUPAC Name 4-nitrobenzoyl chloride
SMILES [O-][N+](=O)C1=CC=C(C=C1)C(Cl)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 70°C to 74°C
Color Yellow
Boiling Point 202°C to 205°C (105.0 mmHg)
Flash Point 102°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula O2NC6H4COCl
Quantity 250 g
Beilstein 09, 394
Fieser 01,736
Merck Index 15, 6677
Solubility Information Solubility in water: decomposes in water. Other solubilities: decomposes in alcohol,soluble in diethyl ether
Formula Weight 185.57
Percent Purity 98%
Physical Form Crystals or Flakes
Chemical Name or Material 4-Nitrobenzoyl chloride, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 204-517-4
RTECSNumber DM6651000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.