Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Valeric anhydride

Catalog No. 245933100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-593-3100ML 100 mL
24-593-3500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 24-593-3100ML Supplier Sigma Aldrich Supplier No. 245933100ML
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Valeric anhydride

Specifications

Melting Point -56°C (lit.)
CAS 2082-59-9
Boiling Point 228°C to 230°C (lit.)
Molecular Formula C10H18O3
Linear Formula [CH3(CH2)3 CO]2 O
MDL Number MFCD00009477
Synonym Pentanoic anhydride
Molecular Weight (g/mol) 186.25
Percent Purity 97%
Density 0.944 g/mL (at 20°C (literature))
Quantity 100 mL
Refractive Index n20/D 1.421 (literature)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.