Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

delta-Valerolactone, 99%

Catalog No. p-211298
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC162710500 50 g Glass bottle
AC162712500 250 g Glass bottle
AC162710010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC162710500 Supplier Thermo Scientific Chemicals Supplier No. 162710500
Only null left
Add to Cart

CAS: 542-28-9 | C5H8O2 | 100.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 542-28-9
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00006645
InChI Key OZJPLYNZGCXSJM-UHFFFAOYSA-N
Synonym delta-valerolactone, tetrahydro-2h-pyran-2-one, 5-valerolactone, 2h-pyran-2-one, tetrahydro, tetrahydropyran-2-one, tetrahydro-2-pyranone, penta-1,5-lactone, .delta.-valerolactone, .delta.-valeryllactone, tetrahydro-pyran-2-one
PubChem CID 10953
ChEBI CHEBI:16545
IUPAC Name oxan-2-one
SMILES O=C1CCCCO1

Specifications

Melting Point -13.0°C
Color Colorless to Yellow
Density 1.1000g/mL
Boiling Point 230.0°C
Flash Point 112°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4560 to 1.4590
Quantity 50 g
Beilstein 17, 235
Specific Gravity 1.1
Solubility Information Solubility in water: miscible
Formula Weight 100.12
Percent Purity 99%
Concentration or Composition (by Analyte or Components) May contain variable amounts of polymerized product
Physical Form Liquid
Chemical Name or Material δ-Valerolactone, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Precautionary Statement GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 208-807-1
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.