Learn More
Cayman Chemical Valeroyl SalIcylate 50mg
SDP
Catalog No. 502895172
Encompass_Preferred
Valeroyl salicylate is a selective, irreversible inhibitor of COX-1. The IC50 values for ovine COX-1 and -2 are 0.8 mM and 15 mM, respectively.{1364} The half-lives for inactivation of human recombinant and ovine COX-1 in the presence of 500 µM valeroyl salicylate are 12 and 45 minutes, respectively.{1284,1364}
Catalog No. 50-289-5172 Supplier Cayman Chemical Supplier No. 7067050 MG
Valeroyl salicylate is a selective, irreversible inhibitor of COX-1. The IC50 values for ovine COX-1 and -2 are 0.8 mM and 15 mM, respectively.{1364} The half-lives for inactivation of human recombinant and ovine COX-1 in the presence of 500 µM valeroyl salicylate are 12 and 45 minutes, respectively.{1284,1364}