Learn More
Medchemexpress LLC Valerylcarnitine | 40225-14-7 | ≥99.0% | 245.32 | 100 MG
SDP
Catalog No. 5000436023
Encompass_Preferred
Valerylcarnitine is an endogenous metabolite belonging to the short-chain acylcarnitines. It is also identified as a Human Endogenous Metabolite. This product is intended for research use only.
  • Available as a solid
  • White to off-white solid
  • Stable as a powder for 3 years at -20°C
  • Stable in solvent for 6 months at -80°C or 1 month at -20°C
Catalog No. 50-004-36023 Supplier Medchemexpress LLC Supplier No. HY113266100MG
Valerylcarnitine is an endogenous metabolite belonging to the short-chain acylcarnitines. It is also identified as a Human Endogenous Metabolite. This product is intended for research use only.
  • Available as a solid
  • White to off-white solid
  • Stable as a powder for 3 years at -20°C
  • Stable in solvent for 6 months at -80°C or 1 month at -20°C