Learn More
Cayman Chemical Valeryl-L-carnitine (chloride), Purity: ≥98%, CAS: 162040-64-4, 50 mg
SDP
Catalog No. 502933609
Encompass_Preferred
A short-chain acylcarnitine; levels increase in the serum of rhesus monkeys following exposure to 7 and 10 Gy of ionizing radiation
Catalog No. 50-293-3609 Supplier Cayman Chemical Supplier No. 2656350 MG
A short-chain acylcarnitine; levels increase in the serum of rhesus monkeys following exposure to 7 and 10 Gy of ionizing radiation