Learn More
Medchemexpress LLC Valganciclovir hydrochloride | 175865-59-5 | 98.0% | C14H23ClN6O5 | 10 MM / 1 ML
SDP
Catalog No. 5000433504
Encompass_Preferred
Valganciclovir hydrochloride is an orally active antiviral agent that inhibits the growth of adenoviruses and offers a protective effect on immunosuppressed hamsters. It is also used for Cytomegalovirus research.
  • Inhibits the growth of human adenoviruses
  • Offers a protective effect on immunosuppressed hamsters
  • Used for Cytomegalovirus research
Catalog No. 50-004-33504 Supplier Medchemexpress LLC Supplier No. HYA0032A10MM/1ML
Valganciclovir hydrochloride is an orally active antiviral agent that inhibits the growth of adenoviruses and offers a protective effect on immunosuppressed hamsters. It is also used for Cytomegalovirus research.
  • Inhibits the growth of human adenoviruses
  • Offers a protective effect on immunosuppressed hamsters
  • Used for Cytomegalovirus research