Learn More
Cayman Chemical Valganciclovir (hydrochloride), Purity: ≥95%, CAS: 175865-59-5, 10 mg
SDP
Catalog No. 502955323
Encompass_Preferred
A prodrug form of ganciclovir; increases survival and reduces weight loss in an immunosuppressed hamster model of disseminated Ad5 infection at 200 mg/kg
Catalog No. 50-295-5323 Supplier Cayman Chemical Supplier No. 1387510 MG
A prodrug form of ganciclovir; increases survival and reduces weight loss in an immunosuppressed hamster model of disseminated Ad5 infection at 200 mg/kg