Learn More
Cayman Chemical ValgncIclovIrhydroCLOrID 100mg
SDP
Catalog No. 502954354
Encompass_Preferred
A prodrug form of ganciclovir; increases survival and reduces weight loss in an immunosuppressed hamster model of disseminated Ad5 infection at 200 mg/kg
Catalog No. 50-295-4354 Supplier Cayman Chemical Supplier No. 13875100 MG
A prodrug form of ganciclovir; increases survival and reduces weight loss in an immunosuppressed hamster model of disseminated Ad5 infection at 200 mg/kg