Learn More
ATHENA ES VALI Medium
Catalog No. 5000158144
Encompass_Preferred
VALI (Vertex modified Air-Liquid Interface) medium is derived from ALI medium. The VALI version was developed to support drug-discovery using human bronchial epithelial cells in an air-liquid surface model
Catalog No. 50-001-58144 Supplier ATHENA ES Supplier No. 0450
VALI (Vertex modified Air-Liquid Interface) medium is derived from ALI medium. The VALI version was developed to support drug-discovery using human bronchial epithelial cells in an air-liquid surface model