Learn More
Medchemexpress LLC HY-B0856 100mg Medchemexpress, Validamycin A CAS:37248-47-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0856100MG
Encompass_Preferred
Medchemexpress, HY-B0856 100mg Validamycin A CAS:37248-47-8 Validamycin A is an aminoglycoside agricultural antibiotic. Validamycin A inhibits the growth of A. flavus, with a MIC of 1 μg/mL. Validamycin A is a reversible tyrosinase inhibitor, with a Ki of 5.893 mM[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1135