Learn More
Research Products International Corp VALIDAMYCIN A 1GM
SDP
Catalog No. 50213729
Encompass_Preferred
Validamycin A is an antifungal compound.  It is a naturally occurring aminocyclitol glycoside and belongs to the class of compounds known as validamycins. 
The primary mode of action of validamycin A involves inhibiting trehalase, an enzyme responsible for breaking down trehalose, a type of sugar. Trehalose is important for fungal cell wall integrity and function. By interfering with trehalose metabolism, validamycin A disrupts the fungal cell structure and inhibits its ability to thrive, ultimately leading to the suppression of fungal growth. 
Catalog No. 50-213-729 Supplier Research Products International Corp Supplier No. V210201.0
Validamycin A is an antifungal compound.  It is a naturally occurring aminocyclitol glycoside and belongs to the class of compounds known as validamycins. 
The primary mode of action of validamycin A involves inhibiting trehalase, an enzyme responsible for breaking down trehalose, a type of sugar. Trehalose is important for fungal cell wall integrity and function. By interfering with trehalose metabolism, validamycin A disrupts the fungal cell structure and inhibits its ability to thrive, ultimately leading to the suppression of fungal growth. 