Learn More
Cayman Chemical ValIdamycIn A 500mg
SDP
Catalog No. 502971758
Encompass_Preferred
A fungicidal agent that inhibits trehalase, a trehalose-hydrolizing enzyme (Ki = 1.9 nM; IC50 = 0.7 μM); used to protect rice plants against sheath blight caused by the pathogenic fungus, R. solani and also effective against the growth and sporulation of R. cerealis, F. culmorum, and other fungi
Catalog No. 50-297-1758 Supplier Cayman Chemical Supplier No. 15326500 MG
A fungicidal agent that inhibits trehalase, a trehalose-hydrolizing enzyme (Ki = 1.9 nM; IC50 = 0.7 μM); used to protect rice plants against sheath blight caused by the pathogenic fungus, R. solani and also effective against the growth and sporulation of R. cerealis, F. culmorum, and other fungi