Learn More
Cayman Chemical Validamycin A, Purity: ≥60%, CAS: 37248-47-8, 5 g
SDP
Catalog No. 502972261
Encompass_Preferred
A fungicidal agent that inhibits trehalase, a trehalose-hydrolizing enzyme (Ki = 1.9 nM; IC50 = 0.7 μM); used to protect rice plants against sheath blight caused by the pathogenic fungus, R. solani and also effective against the growth and sporulation of R. cerealis, F. culmorum, and other fungi
Catalog No. 50-297-2261 Supplier Cayman Chemical Supplier No. 153265 G
A fungicidal agent that inhibits trehalase, a trehalose-hydrolizing enzyme (Ki = 1.9 nM; IC50 = 0.7 μM); used to protect rice plants against sheath blight caused by the pathogenic fungus, R. solani and also effective against the growth and sporulation of R. cerealis, F. culmorum, and other fungi