Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Validamycin A, G-Biosciences
SCI_ED

Catalog No. S139743
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139743 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139743 Supplier G-Biosciences Supplier No. RC1128
Add to Cart
Add to Cart

Validamycin is a fungicidal, aminoglycoside antibiotic known to inhibit trehalases in plants, insects, and fungi as well as enhance trehalose accumulation in transgenic plants.

It is effective against Pellicularia sasakii and Rhizoctonia solani in plants, but only decreases their virulence instead of exhibiting a fungicidal effect. In addition, validamycin is used to control sheath-blight disease in rice and is most effective against soil-borne pathogens.

Chemical Identifiers

Molecular Weight (g/mol) 497.49 g/mol

Specifications

Boiling Point 813.66°C
Quantity 1 g
Percent Purity >60%
Grade Molecular Biology
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.