Learn More
Thermo Scientific™ Validation Tools for UV-Visible Spectrophotometers
Catalog No. 14380470 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
GENESYS Validator
Performance Verification Kit
Qualification Filter Kit
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
14-380-470 GENESYS Validator
14-380-108 Qualification Filter Kit
14-380-109 Performance Verification Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-380-470 Supplier Thermo Scientific™ Supplier No. 840323100
Add to Cart
Add to Cart
Ensure compliance with regulatory guidelines with the Thermo Scientific™ Validation Tools for UV-Visible Spectrophotometers providing tools and documentation for system qualification.
Get all of the documentation and reference materials needed to facilitate Installation and Operational qualification procedures (IQ/OQ) with Thermo Scientific™ Validation Tools for UV-Visible Spectrophotometers. Certified standards for verifying instrument performance to factory specifications are available separately.
UV-Visible Spectrophotometer Validation Solutions include:
  • Information on the validation process
  • Documentation for manufacturer-verified Installation Qualification (IQ) to ensure proper system performance
  • Information on pharmacopoeia recommended tests for specific instrument performance where applicable
  • Installation and instrument certification
Solutions available for GENESYS 40-180, BioMate 160, Evolution200 series, Evolution 350, NanoDrop One, NanoDrop 2000, and NanoDrop 8000 instruments
TRUSTED_SUSTAINABILITY
Specifications
Type GENESYS Validator
For Use With (Equipment) GENESYS 40-180, BioMate 160 Spectrophotometers