Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Valiglurax | 1976050-09-5 | ≥98.0% | 25 MG
SDP

Catalog No. 5000230637
Encompass_Preferred

Valiglurax is a potent, orally active, and selective positive allosteric modulator of the metabotropic glutamate receptor mGlu4 (VU0652957). It is CNS-penetrant and intended for preclinical research applications such as Parkinson's disease models.

  • Potent mGlu4 positive allosteric modulator with low-nanomolar activity.
  • Orally active and central nervous system penetrant.
  • Solid, light yellow to yellow, suitable for research use.
  • Molecular formula C16H10F3N5; molecular weight 329.28 g/mol.
  • Storage: 4°C, protect from light; in solvent: -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-30637 Supplier Medchemexpress LLC Supplier No. HY12264725MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Valiglurax is a potent, orally active, and selective positive allosteric modulator of the metabotropic glutamate receptor mGlu4 (VU0652957). It is CNS-penetrant and intended for preclinical research applications such as Parkinson's disease models.

  • Potent mGlu4 positive allosteric modulator with low-nanomolar activity.
  • Orally active and central nervous system penetrant.
  • Solid, light yellow to yellow, suitable for research use.
  • Molecular formula C16H10F3N5; molecular weight 329.28 g/mol.
  • Storage: 4°C, protect from light; in solvent: -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.