Learn More
Medchemexpress LLC Valiltramiprosate | 100MG
SDP
Catalog No. 5000227649
Encompass_Preferred
ALZ-801 is a potent and orally available small-molecule -amyloid (A ) anti-oligomer and aggregation inhibitor valine-conjugated proagent of Tramiprosate with substantially improved PK properties and gastrointestinal tolerability compared with the parent compound[1] ALZ-801 is an advanced and markedly improved candidate for the treatment of alzheimer s disease[2]
Catalog No. 50-002-27649 Supplier Medchemexpress LLC Supplier No. HY117259100MG
ALZ-801 is a potent and orally available small-molecule -amyloid (A ) anti-oligomer and aggregation inhibitor valine-conjugated proagent of Tramiprosate with substantially improved PK properties and gastrointestinal tolerability compared with the parent compound[1] ALZ-801 is an advanced and markedly improved candidate for the treatment of alzheimer s disease[2]