Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Valiltramiprosate 10mg | 1034190-08-3 | 10 MG
SDP

Supplier:  Medchemexpress LLC HY11725910MG

Encompass_Preferred

Valiltramiprosate is an orally active valine prodrug of tramiprosate developed as a small-molecule inhibitor of β-amyloid (Aβ) oligomer formation for Alzheimer's disease research. It is supplied as a solid for use in in vitro and preclinical studies.

  • Valine prodrug of tramiprosate.
  • Inhibits β-amyloid oligomer formation.
  • Supplied as solid in 10 mg packs.
  • Purity 98.8%.
  • Molecular formula C8H18N2O4S; molecular weight 238.30 g/mol.
  • For research use only.

Catalog No. 50-002-26473


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.