Learn More
Medchemexpress LLC Valiltramiprosate 25mg | 1034190-08-3 | 25 MG
SDP
Catalog No. 5000227741
Encompass_Preferred
Valiltramiprosate is an orally available small-molecule inhibitor of β-amyloid oligomer formation and aggregation supplied for laboratory research. The compound is provided as a solid 25 mg unit with high purity and is intended for research use only; store sealed and protected from moisture.
  • Orally available small-molecule inhibitor of β-amyloid aggregation.
  • Supplied as a 25 mg solid for preclinical and in vitro studies.
  • High purity (≈98.8%) suitable for research applications.
  • Molecular weight 238.30 g·mol⁻¹ and formula C8H18N2O4S.
  • Store sealed at recommended temperatures to preserve stability.
Catalog No. 50-002-27741 Supplier Medchemexpress LLC Supplier No. HY11725925MG
Valiltramiprosate is an orally available small-molecule inhibitor of β-amyloid oligomer formation and aggregation supplied for laboratory research. The compound is provided as a solid 25 mg unit with high purity and is intended for research use only; store sealed and protected from moisture.
  • Orally available small-molecule inhibitor of β-amyloid aggregation.
  • Supplied as a 25 mg solid for preclinical and in vitro studies.
  • High purity (≈98.8%) suitable for research applications.
  • Molecular weight 238.30 g·mol⁻¹ and formula C8H18N2O4S.
  • Store sealed at recommended temperatures to preserve stability.