Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Valiltramiprosate 50mg | 1034190-08-3 | 238.30 | C8H18N2O4S | 50 MG
SDP

Catalog No. 5000227798
Encompass_Preferred

Valiltramiprosate (ALZ-801) is a valine-conjugated prodrug of tramiprosate developed as an inhibitor of β-amyloid oligomer formation for research use. The compound has CAS 1034190-08-3, molecular formula C8H18N2O4S, molecular weight ≈238.3 g/mol, and a COA-reported purity of 98.83% (LCMS). Store sealed, away from moisture; in solvent, -80°C up to 6 months or -20°C up to 1 month.

  • Valine-conjugated prodrug of tramiprosate
  • Inhibits β-amyloid oligomer formation
  • CAS 1034190-08-3 and molecular formula C8H18N2O4S
  • Purity 98.83% by LCMS
  • Storage: sealed, away from moisture; in solvent -80°C (6 months) or -20°C (1 month)

Catalog No. 50-002-27798 Supplier Medchemexpress LLC Supplier No. HY11725950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Valiltramiprosate (ALZ-801) is a valine-conjugated prodrug of tramiprosate developed as an inhibitor of β-amyloid oligomer formation for research use. The compound has CAS 1034190-08-3, molecular formula C8H18N2O4S, molecular weight ≈238.3 g/mol, and a COA-reported purity of 98.83% (LCMS). Store sealed, away from moisture; in solvent, -80°C up to 6 months or -20°C up to 1 month.

  • Valine-conjugated prodrug of tramiprosate
  • Inhibits β-amyloid oligomer formation
  • CAS 1034190-08-3 and molecular formula C8H18N2O4S
  • Purity 98.83% by LCMS
  • Storage: sealed, away from moisture; in solvent -80°C (6 months) or -20°C (1 month)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.