Learn More
Medchemexpress LLC Valiltramiprosate 5mg | 1034190-08-3 | 238.30 g/mol | C8H18N2O4S | 5 MG
SDP
Catalog No. 5000227823
Encompass_Preferred
Valiltramiprosate (also known as ALZ-801) is a valine-conjugated prodrug of tramiprosate that inhibits β-amyloid aggregation and oligomer formation. Supplied for research use only, it is intended for preclinical and in vitro studies exploring amyloid-β pathology.
  • Inhibits β-amyloid aggregation and oligomer formation.
  • Valine prodrug design improves pharmacokinetic and tolerability profile relative to tramiprosate.
  • Suitable for in vitro and preclinical assays requiring small-scale compound quantities.
  • High purity (98.83% by LCMS) for research applications.
  • Well-defined chemical identity for analytical and formulation work.
Catalog No. 50-002-27823 Supplier Medchemexpress LLC Supplier No. HY1172595MG
Valiltramiprosate (also known as ALZ-801) is a valine-conjugated prodrug of tramiprosate that inhibits β-amyloid aggregation and oligomer formation. Supplied for research use only, it is intended for preclinical and in vitro studies exploring amyloid-β pathology.
  • Inhibits β-amyloid aggregation and oligomer formation.
  • Valine prodrug design improves pharmacokinetic and tolerability profile relative to tramiprosate.
  • Suitable for in vitro and preclinical assays requiring small-scale compound quantities.
  • High purity (98.83% by LCMS) for research applications.
  • Well-defined chemical identity for analytical and formulation work.