Learn More
Medchemexpress LLC Valine-2,3,4,4,4,4',4',4'-d8 | 203784-63-8 | 99.8% | 125.20 | 25 MG
SDP
Catalog No. 5000452710
Encompass_Preferred
DL-Valine-d8 is the deuterium labeled DL-Valine.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-52710 Supplier Medchemexpress LLC Supplier No. 25MGHYW012889S
DL-Valine-d8 is the deuterium labeled DL-Valine.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.