Learn More
Medchemexpress LLC Valine-2,3,4,4,4,4',4',4'-d8 | 203784-63-8 | 99.8% | 125.20 | 5 MG
SDP
Catalog No. 5000454944
Encompass_Preferred
DL-Valine-d8 is the deuterium labeled DL-Valine. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Isotope-labeled compounds
  • Internal standard for clinical mass spectrometry
  • Internal standard for food
  • Isotope-labeled amino acids & peptides
  • Deuterium (2H, D)
  • SILAC reagents
  • Newborn screening
  • Isotope-labeled amino acids
Catalog No. 50-004-54944 Supplier Medchemexpress LLC Supplier No. HYW012889S5MG
DL-Valine-d8 is the deuterium labeled DL-Valine. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Isotope-labeled compounds
  • Internal standard for clinical mass spectrometry
  • Internal standard for food
  • Isotope-labeled amino acids & peptides
  • Deuterium (2H, D)
  • SILAC reagents
  • Newborn screening
  • Isotope-labeled amino acids