Learn More
Medchemexpress LLC Valine-2,3,4,4,4,4',4',4'-d8 | 203784-63-8 | MFCD00144194 | 125.20 | 10 MG
SDP
Catalog No. 5000454881
Encompass_Preferred
DL-Valine-d8 is the deuterium labeled DL-Valine. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-54881 Supplier Medchemexpress LLC Supplier No. HYW012889S10MG
DL-Valine-d8 is the deuterium labeled DL-Valine. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.