Learn More
Cambridge Isotope Laboratories L-Valine (D8, 98%), 35045-72-8, MFCD00144704, 0.25G
Supplier:  Cambridge Isotope Laboratories DLM4880.25
Encompass
Chemical Formula: (CD3)2CDCD(NH2)COOH, Molecular Weight: 125.20, Synonym: (S)-α-Aminoisovaleric acid-2,3,4,4,4,5,5,5-d8, Deuterated L-valine, L-2-Amino-3-methylbutanoic acid-2,3,4,4,4,5,5,5-d8, MP: 295-300 °C (subl.) (lit.)
Catalog No. NC0894143