Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Valine (L), USP
SDP

Catalog No. 14186148 Shop All Medisca Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-186-148 100 g
14-185-128 500 g
14-185-171 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-186-148 Supplier Medisca Supplier No. 035805
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Valine (L), USP comes as white, odorless crystals which are soluble in water and practically insoluble in alcohol.

Valine (L), USP comes as white, odorless crystals which are soluble in water and practically insoluble in alcohol.

Alternative name(s): L-valine, L-val

Specifications

Chemical Name or Material Valine (L), USP
Packaging Jar, 300mL, White, HDPE, Round, 63-400, TE Pano Neck
CAS 72-18-4
Type Drug Standard
Molecular Weight (g/mol) 117.147
Quantity 100 g
Percent Purity 98.5 to 101.5 %
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.