Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Valine, USP, 98.5-101.5%, Spectrum™ Chemical
SDP

Catalog No. 18607683 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-683 1 kg Poly Bottle
18-607-681 100 g Poly Bottle
18-607-682 500 g Poly Bottle
18-607-680 25 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-607-683 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. V10051KG
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

V1005, 72-18-4, C5H11NO2

Spectrum™ Chemical Valine, USP - All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 72-18-4
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00064220
InChI Key KZSNJWFQEVHDMF-BYPYZUCNSA-N
IUPAC Name (2S)-2-amino-3-methylbutanoic acid
SMILES CC(C)[C@H](N)C(O)=O

Specifications

CAS Max % 1
pH 5.5 to 7.0
Ignition Residue 0.001
Loss on Drying 0.003
Packaging Poly Bottle
Quantity 1 kg
Specific Rotation +26.6° to +28.8°
Percent Purity 98.5 to 101.5%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Valine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.