Learn More
Medchemexpress LLC Valinomycin | 2001-95-8 | 50 MG
SDP
Catalog No. 5000417775
Encompass_Preferred
Valinomycin is a potassium-specific ionophore that forms a complex with K+ ions. This complex can incorporate into biological bilayer membranes, disrupting the normal potassium gradient and inducing cell death. When integrated into liposomes, it shows reduced cytotoxicity and improved targeting. It demonstrates a broad range of biological activities, including antibiotic, antifungal, antiviral, antitumor, and insecticidal efficacies, making it valuable for various research applications.
  • Potassium-specific ionophore.
  • Disrupts normal potassium gradients across cell membranes.
  • Reduces cytotoxicity and enhances targeting when incorporated into liposomes.
  • Exhibits antibiotic, antifungal, antiviral, antitumor, and insecticidal properties.
  • Suppresses viability and induces apoptosis in HepG2 cells.
  • Inhibits tumor growth in in vivo models.
Catalog No. 50-004-17775 Supplier Medchemexpress LLC Supplier No. HYN669350MG
Valinomycin is a potassium-specific ionophore that forms a complex with K+ ions. This complex can incorporate into biological bilayer membranes, disrupting the normal potassium gradient and inducing cell death. When integrated into liposomes, it shows reduced cytotoxicity and improved targeting. It demonstrates a broad range of biological activities, including antibiotic, antifungal, antiviral, antitumor, and insecticidal efficacies, making it valuable for various research applications.
  • Potassium-specific ionophore.
  • Disrupts normal potassium gradients across cell membranes.
  • Reduces cytotoxicity and enhances targeting when incorporated into liposomes.
  • Exhibits antibiotic, antifungal, antiviral, antitumor, and insecticidal properties.
  • Suppresses viability and induces apoptosis in HepG2 cells.
  • Inhibits tumor growth in in vivo models.