Learn More
Enzo Life Sciences Valinomycin (25mg). CAS: 2001-95-8
SDP
Catalog No. 502011064
Encompass_Preferred
A potassium ionophore. Induces mitochondrial swelling. Induces apoptosis in CHO cells and other cell lines. Antagonizes endothelin-induced vasoconstriction (IC50=0.3µM). Purity: ≥97% (TLC), ≥92% (HPLC). Solubility: Soluble in 100% ethanol (50mg/ml), acetic acid, chloroform, DMSO or ether. Long Term Storage: -20°C.
Catalog No. 50-201-1064 Supplier Enzo Life Sciences Supplier No. BMLKC1400025
A potassium ionophore. Induces mitochondrial swelling. Induces apoptosis in CHO cells and other cell lines. Antagonizes endothelin-induced vasoconstriction (IC50=0.3µM). Purity: ≥97% (TLC), ≥92% (HPLC). Solubility: Soluble in 100% ethanol (50mg/ml), acetic acid, chloroform, DMSO or ether. Long Term Storage: -20°C.