Learn More
Medchemexpress LLC HY-N6693 5mg Medchemexpress, Valinomycin CAS:2001-95-8 Purity:>98%
SDP
Catalog No. 501970456
Encompass_Preferred
Medchemexpress, HY-N6693 5mg Valinomycin CAS:2001-95-8 Valinomycin (NSC 122023) is a cyclic depsipeptide antibiotic first isolated from Streptomyces fulvissimus, act as a potassium selective ionophore. Valinomycin (NSC 122023) inhibits lymphocyte proliferation by its effects on the cell membrane, and induces apoptosis in CHO cells. Valinomycin induces activation of PINK1 leading to Parkin Ser65 phosphorylation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0456 Supplier Medchemexpress LLC Supplier No. HYN66935MG
Medchemexpress, HY-N6693 5mg Valinomycin CAS:2001-95-8 Valinomycin (NSC 122023) is a cyclic depsipeptide antibiotic first isolated from Streptomyces fulvissimus, act as a potassium selective ionophore. Valinomycin (NSC 122023) inhibits lymphocyte proliferation by its effects on the cell membrane, and induces apoptosis in CHO cells. Valinomycin induces activation of PINK1 leading to Parkin Ser65 phosphorylation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.