Learn More
Sigma Aldrich Fine Chemicals Biosciences Valinomycin 99.0 TLC10MG
Catalog No. 502899339
Encompass
Valinomycinnbspis a neutral cyclic dodecadepsipeptide antibiotic that has a 36-member ring.
Catalog No. 50-289-9339 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 9467510MG
Valinomycinnbspis a neutral cyclic dodecadepsipeptide antibiotic that has a 36-member ring.