Learn More
Sigma Aldrich Fine Chemicals Biosciences Valinomycin 99.0 TLC500MG
Catalog No. 502903354
Encompass
Valinomycinnbspis a neutral cyclic dodecadepsipeptide antibiotic that has a 36-member ring.
Catalog No. 50-290-3354 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 94675500MG
Valinomycinnbspis a neutral cyclic dodecadepsipeptide antibiotic that has a 36-member ring.