Learn More
Cayman Chemical Valinomycin Fatty Acids and De
SDP
Catalog No. 502888578
Encompass_Preferred
A cyclododecadepsipeptide potassium-selective ionophore antibotic; induces apoptosis in several cell tyes, including CHO cells, by stimulating potassium efflux
Catalog No. 50-288-8578 Supplier Cayman Chemical Supplier No. 1000915225 MG
A cyclododecadepsipeptide potassium-selective ionophore antibotic; induces apoptosis in several cell tyes, including CHO cells, by stimulating potassium efflux