Learn More
Alkali Scientific VALINOMYCIN, STREPTOMYCES FULVISSIMUS, 100 Mg
SDP
Catalog No. 5000659644
Encompass_Preferred
Valinomycin, 100mg, is an ion transport inhibitor that plays a critical role in cellular ion regulation and mitochondrial function. This powder form is typically used in laboratory research focused on studying ion channels and transport mechanisms within biological membranes. By forming complexes with potassium ions, Valinomycin facilitates the movement of these ions across cellular membranes, which is crucial in experiments exploring ion channel dynamics and cellular signal transduction. The 100mg format is suited for a wide range of research applications, particularly in studies of ionophores and their effects on ion transport in various experimental systems.
Catalog No. 50-006-59644 Supplier Alkali Scientific Supplier No. INO002CP
Valinomycin, 100mg, is an ion transport inhibitor that plays a critical role in cellular ion regulation and mitochondrial function. This powder form is typically used in laboratory research focused on studying ion channels and transport mechanisms within biological membranes. By forming complexes with potassium ions, Valinomycin facilitates the movement of these ions across cellular membranes, which is crucial in experiments exploring ion channel dynamics and cellular signal transduction. The 100mg format is suited for a wide range of research applications, particularly in studies of ionophores and their effects on ion transport in various experimental systems.