Learn More
Alkali Scientific VALINOMYCIN, STREPTOMYCES FULVISSIMUS, 50 Mg
SDP
Supplier:  Alkali Scientific INO001CP
Encompass_Preferred
Valinomycin, a potent ion transport inhibitor, is a natural product used to study membrane transport processes and mitochondrial function. Available in powder form, the 50mg vial is ideal for laboratory applications requiring small amounts of this powerful ionophore. Valinomycin works by binding potassium ions and facilitating their transport across lipid membranes, making it useful in studies of potassium channel activity and cellular ion homeostasis. It is commonly used in cell biology and pharmacological research, as well as in experiments studying the mechanisms of ion transport and cellular signaling pathways, particularly in the context of ionophores in pharmacology.
Catalog No. 50-006-59701