Learn More
Cayman Chemical Valtrate, Purity: ≥98%, CAS: 18296-44-1, 10 mg
SDP
Catalog No. 502911599
Encompass_Preferred
A valepotriate with diverse biological activities; inhibits nuclear export of influenza vRNP in HeLa cells at 3.6 µM; inhibits influenza virus infection in MDCK cells (IC50 = 0.19 µM); cytotoxic to GLC-4 and COLO 320 cells (IC50s = 1.4 and 3 µM, respectively); increases the number of entries into, and percentage of time spent in, the open arms of the elevated plus maze, indicating anxiolytic-like activity, and decreases serum corticosterone levels in rats at 10 mg/kg
Catalog No. 50-291-1599 Supplier Cayman Chemical Supplier No. 3652210 MG
A valepotriate with diverse biological activities; inhibits nuclear export of influenza vRNP in HeLa cells at 3.6 µM; inhibits influenza virus infection in MDCK cells (IC50 = 0.19 µM); cytotoxic to GLC-4 and COLO 320 cells (IC50s = 1.4 and 3 µM, respectively); increases the number of entries into, and percentage of time spent in, the open arms of the elevated plus maze, indicating anxiolytic-like activity, and decreases serum corticosterone levels in rats at 10 mg/kg