Learn More
Selleck Chemical LLC Valnemulin HCl S4216-50mg
SDP
Catalog No. 503111115
Encompass_Preferred
Valnemulin HCl is a broad-spectrum bacteriostatic agent inhibiting protein synthesis in bacteria by binding to the peptidyl transferase component of the 50S subunit of ribosomes
Catalog No. 50-311-1115 Supplier Selleck Chemical LLC Supplier No. S421650MG
Add to Cart
Add to Cart
Valnemulin HCl is a broad-spectrum bacteriostatic agent inhibiting protein synthesis in bacteria by binding to the peptidyl transferase component of the 50S subunit of ribosomes