Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Valnemulin HCl, G-Biosciences
SCI_ED

Catalog No. S157236
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S157236 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S157236 Supplier G-Biosciences Supplier No. RC1129
Add to Cart
Add to Cart

Valnemulin is a pleuromutilin antibiotic targeting Mycoplasma bacteria, especially those responsible for bovine respiratory infections.

The mechanism of action of Valnemulin involves its inhibition of protein synthesis in bacteria by binding the peptidyl transferase enzyme in the 50s ribosomal subunit.

Chemical Identifiers

Molecular Weight (g/mol) 601.28 g/mol

Specifications

Boiling Point 672.7°C
Quantity 500 mg
Percent Purity 98%
Grade Molecular Biology
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.