Learn More
Medchemexpress LLC Valnemulin hydrochloride | 133868-46-9 | 98.1% | C31H53ClN2O5S | 1 ML
SDP
Catalog No. 5000432052
Encompass_Preferred
Valnemulin hydrochloride is an orally effective truncated pleurotin antibiotic. It inhibits bacterial protein synthesis by binding to peptidyl transferase in the 50s ribosome subunit. This antibiotic has shown efficacy in eliminating Mycobacterium bovis in experimental bovine models of Mycoplasma bovis infection and can reduce mortality in epidemic rabbit enteropathy.
  • Inhibits bacterial protein synthesis
  • Effectively eliminates Mycobacterium bovis in bovine models
  • Reduces mortality in epidemic rabbit enteropathy
  • Supports rapid diminution of clinical signs and restoration of appetite in calves
  • No adverse effects on rabbit growth performance
Catalog No. 50-004-32052 Supplier Medchemexpress LLC Supplier No. HYB002710MM/1ML
Valnemulin hydrochloride is an orally effective truncated pleurotin antibiotic. It inhibits bacterial protein synthesis by binding to peptidyl transferase in the 50s ribosome subunit. This antibiotic has shown efficacy in eliminating Mycobacterium bovis in experimental bovine models of Mycoplasma bovis infection and can reduce mortality in epidemic rabbit enteropathy.
  • Inhibits bacterial protein synthesis
  • Effectively eliminates Mycobacterium bovis in bovine models
  • Reduces mortality in epidemic rabbit enteropathy
  • Supports rapid diminution of clinical signs and restoration of appetite in calves
  • No adverse effects on rabbit growth performance