Learn More
Medchemexpress LLC Valnivudine hydrochloride | 956483-03-7 | 98.99% | 534.04 | 50 MG
SDP
Catalog No. 5000401728
Encompass_Preferred
Valnivudine hydrochloride (FV-100) is a potent, selective, and orally active anti-varicella zoster virus agent. It is a proagent of CF-1743 and exhibits very low toxicity in vivo. It is intended for research use only.
  • Potent, selective, and orally active anti-varicella zoster virus agent.
  • Proagent of CF-1743.
  • Exhibits very low toxicity in vivo.
  • Intensely fluorescent when illuminated by wavelengths of 340-380 nm (In Vitro).
  • Does not induce biologically relevant respiratory changes or apparent neuropharmacological effects in rats at 50-500 mg/kg; p.o. (In Vivo).
Catalog No. 50-004-01728 Supplier Medchemexpress LLC Supplier No. HY1985650MG
Valnivudine hydrochloride (FV-100) is a potent, selective, and orally active anti-varicella zoster virus agent. It is a proagent of CF-1743 and exhibits very low toxicity in vivo. It is intended for research use only.
  • Potent, selective, and orally active anti-varicella zoster virus agent.
  • Proagent of CF-1743.
  • Exhibits very low toxicity in vivo.
  • Intensely fluorescent when illuminated by wavelengths of 340-380 nm (In Vitro).
  • Does not induce biologically relevant respiratory changes or apparent neuropharmacological effects in rats at 50-500 mg/kg; p.o. (In Vivo).