Learn More
Medchemexpress LLC Valnivudine hydrochloride | 956483-03-7 | 99.0% | 534.04 | 25 MG
SDP
Catalog No. 5000402205
Encompass_Preferred
Valnivudine hydrochloride (FV-100) is a potent, selective, and orally active anti-varicella zoster virus agent. It is a proagent of CF-1743 and exhibits very low toxicity in vivo. It is intensely fluorescent when illuminated by wavelengths of 340-380 nm and produces a different distribution in HeLa cells within 1 hour. In rats, Valnivudine hydrochloride (50-500 mg/kg; p.o.) does not induce biologically relevant respiratory changes or any apparent neuropharmacological effects.
  • Potent, selective, and orally active anti-varicella zoster virus agent
  • Proagent of CF-1743
  • Exhibits very low toxicity in vivo
  • Intensely fluorescent when illuminated by wavelengths of 340-380 nm
  • Produces a different distribution in HeLa cells within 1 hour
  • Does not induce biologically relevant respiratory changes or apparent neuropharmacological effects in rats
Catalog No. 50-004-02205 Supplier Medchemexpress LLC Supplier No. 25MGHY19856
Valnivudine hydrochloride (FV-100) is a potent, selective, and orally active anti-varicella zoster virus agent. It is a proagent of CF-1743 and exhibits very low toxicity in vivo. It is intensely fluorescent when illuminated by wavelengths of 340-380 nm and produces a different distribution in HeLa cells within 1 hour. In rats, Valnivudine hydrochloride (50-500 mg/kg; p.o.) does not induce biologically relevant respiratory changes or any apparent neuropharmacological effects.
  • Potent, selective, and orally active anti-varicella zoster virus agent
  • Proagent of CF-1743
  • Exhibits very low toxicity in vivo
  • Intensely fluorescent when illuminated by wavelengths of 340-380 nm
  • Produces a different distribution in HeLa cells within 1 hour
  • Does not induce biologically relevant respiratory changes or apparent neuropharmacological effects in rats