Learn More
Medchemexpress LLC Valnivudine hydrochloride | 956483-03-7 | 99.0% | 534.04 | 5 MG
SDP
Catalog No. 5000401734
Encompass_Preferred
FV-100 is a potent, selective, and orally active anti-varicella zoster virus agent. It is a proagent of CF-1743 and demonstrates very low toxicity in vivo.
  • Potent and selective anti-varicella zoster virus (VZV) agent.
  • Orally active.
  • Exhibits very low toxicity in vivo.
  • Intensely fluorescent when illuminated by wavelengths of 340-380 nm.
Catalog No. 50-004-01734 Supplier Medchemexpress LLC Supplier No. HY198565MG
FV-100 is a potent, selective, and orally active anti-varicella zoster virus agent. It is a proagent of CF-1743 and demonstrates very low toxicity in vivo.
  • Potent and selective anti-varicella zoster virus (VZV) agent.
  • Orally active.
  • Exhibits very low toxicity in vivo.
  • Intensely fluorescent when illuminated by wavelengths of 340-380 nm.