Learn More
Medchemexpress LLC Valnivudine hydrochloride | 956483-03-7 | MFCD30146049 | 99.0% | 534.04 g/mol | C27H36ClN3O6 | 10 MG
SDP
Catalog No. 5000296762
Encompass_Preferred
Valnivudine hydrochloride is an orally active nucleoside prodrug of CF-1743 developed as a selective inhibitor of varicella-zoster virus (VZV). It demonstrates potent antiviral activity with low in vivo toxicity and is supplied as a high-purity white to off-white solid for research use, with documented solubility and formulation guidance for in vitro and in vivo studies.
  • Potent, selective VZV inhibition.
  • Orally active prodrug of CF-1743.
  • High purity solid suitable for biochemical and pharmacology studies.
  • Documented in vitro solubility and in vivo formulation protocols.
  • Stable when stored sealed and away from moisture; solvent storage guidance available.
  • Supplied for research use only.
Catalog No. 50-002-96762 Supplier Medchemexpress LLC Supplier No. HY1985610MG
Valnivudine hydrochloride is an orally active nucleoside prodrug of CF-1743 developed as a selective inhibitor of varicella-zoster virus (VZV). It demonstrates potent antiviral activity with low in vivo toxicity and is supplied as a high-purity white to off-white solid for research use, with documented solubility and formulation guidance for in vitro and in vivo studies.
  • Potent, selective VZV inhibition.
  • Orally active prodrug of CF-1743.
  • High purity solid suitable for biochemical and pharmacology studies.
  • Documented in vitro solubility and in vivo formulation protocols.
  • Stable when stored sealed and away from moisture; solvent storage guidance available.
  • Supplied for research use only.