Learn More
Cayman Chemical Valnoctamide, Purity: ≥95%, CAS: 4171-13-5, 100 mg
SDP
Catalog No. 502965197
Encompass_Preferred
An isomer of valpromide that has been marketed as an anxiolytic and sedative compound; abolishes the activity of MIP synthase in human brain crude homogenates (Ki = 0.18 mM); suppresses electrographic seizures in animal models of status epilepticus
Catalog No. 50-296-5197 Supplier Cayman Chemical Supplier No. 18456100 MG
An isomer of valpromide that has been marketed as an anxiolytic and sedative compound; abolishes the activity of MIP synthase in human brain crude homogenates (Ki = 0.18 mM); suppresses electrographic seizures in animal models of status epilepticus