Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Valopicitabine dihydrochloride | 640725-71-9 | MFCD34589916 | >99.0% | 429.3 g/mol | C15H26Cl2N4O6 | 10 MG
SDP

Catalog No. 5000243074
Encompass_Preferred

Valopicitabine dihydrochloride is the dihydrochloride salt of valopicitabine, an orally bioavailable nucleoside prodrug of 2'-C-methylcytidine (NM107) developed as an NS5B polymerase inhibitor for hepatitis C virus. It is provided as a purified powder for research and antiviral biochemical studies.

  • Acts as a nucleoside prodrug of 2'-C-methylcytidine (NM107)
  • Inhibits HCV NS5B polymerase activity in antiviral assays
  • Supplied as a dihydrochloride salt with formula C15H26Cl2N4O6
  • Molecular weight approximately 429.3 g/mol
  • Typical analytical purity ≥99.0% for research use
  • Provided as a powder suitable for biochemical and cellular studies

Catalog No. 50-002-43074 Supplier Medchemexpress LLC Supplier No. HY108060A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Valopicitabine dihydrochloride is the dihydrochloride salt of valopicitabine, an orally bioavailable nucleoside prodrug of 2'-C-methylcytidine (NM107) developed as an NS5B polymerase inhibitor for hepatitis C virus. It is provided as a purified powder for research and antiviral biochemical studies.

  • Acts as a nucleoside prodrug of 2'-C-methylcytidine (NM107)
  • Inhibits HCV NS5B polymerase activity in antiviral assays
  • Supplied as a dihydrochloride salt with formula C15H26Cl2N4O6
  • Molecular weight approximately 429.3 g/mol
  • Typical analytical purity ≥99.0% for research use
  • Provided as a powder suitable for biochemical and cellular studies

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.