Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Valopicitabine (dihydrochloride) | 640725-71-9 | MFCD34589916 | 99.8% | 429.3 g/mol | C15H26Cl2N4O6 | 50 MG
SDP

Catalog No. 5000243087
Encompass_Preferred

Valopicitabine (dihydrochloride) is the dihydrochloride salt of valopicitabine (NM-283), a nucleoside-analog prodrug of 2'-C-methylcytidine with activity against hepatitis C virus through inhibition of the NS5B RNA polymerase. It is provided as a research reagent for biochemical and antiviral studies.

  • High purity suitable for analytical and biological assays.
  • Dihydrochloride salt form, improving aqueous solubility compared with the free base.
  • Acts as an orally bioavailable prodrug of an HCV polymerase inhibitor.
  • Molecular weight 429.3 g/mol; formula C15H26Cl2N4O6.
  • Supplied in small research quantities (e.g., 50 MG) for laboratory use.
  • Accompanied by analytical documentation such as COA, H NMR, and LC-MS.

Catalog No. 50-002-43087 Supplier Medchemexpress LLC Supplier No. HY108060A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Valopicitabine (dihydrochloride) is the dihydrochloride salt of valopicitabine (NM-283), a nucleoside-analog prodrug of 2'-C-methylcytidine with activity against hepatitis C virus through inhibition of the NS5B RNA polymerase. It is provided as a research reagent for biochemical and antiviral studies.

  • High purity suitable for analytical and biological assays.
  • Dihydrochloride salt form, improving aqueous solubility compared with the free base.
  • Acts as an orally bioavailable prodrug of an HCV polymerase inhibitor.
  • Molecular weight 429.3 g/mol; formula C15H26Cl2N4O6.
  • Supplied in small research quantities (e.g., 50 MG) for laboratory use.
  • Accompanied by analytical documentation such as COA, H NMR, and LC-MS.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.