Learn More
Medchemexpress LLC Valopicitabine dihydrochloride | 640725-71-9 | MFCD34589916 | 99.8% | 429.30 g·mol⁻¹ | C15H26Cl2N4O6 | 25 MG
SDP
Catalog No. 5000243003
Encompass_Preferred
Valopicitabine dihydrochloride is the 3-O-valine ester prodrug of the nucleoside analog 2′-C-methylcytidine with antiviral activity against hepatitis C virus (HCV). Supplied as the dihydrochloride salt for research use, it is used in mechanism-of-action studies, preclinical pharmacology, and formulation development.
  • High purity (99.8%) suitable for analytical and preclinical studies.
  • Dihydrochloride salt form improves aqueous solubility for formulation work.
  • Prodrug of 2′-C-methylcytidine enabling studies of prodrug activation.
  • Available in small research quantities for assay development and screening.
  • Characterized by CAS 640725-71-9 and molecular weight 429.30 g·mol⁻¹.
Catalog No. 50-002-43003 Supplier Medchemexpress LLC Supplier No. HY108060A25MG
Valopicitabine dihydrochloride is the 3-O-valine ester prodrug of the nucleoside analog 2′-C-methylcytidine with antiviral activity against hepatitis C virus (HCV). Supplied as the dihydrochloride salt for research use, it is used in mechanism-of-action studies, preclinical pharmacology, and formulation development.
  • High purity (99.8%) suitable for analytical and preclinical studies.
  • Dihydrochloride salt form improves aqueous solubility for formulation work.
  • Prodrug of 2′-C-methylcytidine enabling studies of prodrug activation.
  • Available in small research quantities for assay development and screening.
  • Characterized by CAS 640725-71-9 and molecular weight 429.30 g·mol⁻¹.