Learn More
Medchemexpress LLC Valproic acid | 99-66-1 | 99.7% | 144.21 | 250 MG
SDP
Catalog No. 5000347079
Encompass_Preferred
Valproic acid (VPA) is an orally active HDAC inhibitor. It inhibits HDAC1 and induces proteasomal degradation of HDAC2, while also activating Notch1 signaling and inhibiting proliferation in small cell lung cancer (SCLC) cells. This compound is utilized in various applications.
  • Orally active HDAC inhibitor
  • Inhibits HDAC1 and promotes proteasomal degradation of HDAC2
  • Activates Notch1 signaling
  • Inhibits proliferation in small cell lung cancer (SCLC) cells
  • Used in epilepsy
  • Used in bipolar disorder
  • Used in metabolic disease
  • Used in HIV infection
  • Used for the prevention of migraine headaches
Catalog No. 50-003-47079 Supplier Medchemexpress LLC Supplier No. 25GHY10585
Add to Cart
Add to Cart
Valproic acid (VPA) is an orally active HDAC inhibitor. It inhibits HDAC1 and induces proteasomal degradation of HDAC2, while also activating Notch1 signaling and inhibiting proliferation in small cell lung cancer (SCLC) cells. This compound is utilized in various applications.
  • Orally active HDAC inhibitor
  • Inhibits HDAC1 and promotes proteasomal degradation of HDAC2
  • Activates Notch1 signaling
  • Inhibits proliferation in small cell lung cancer (SCLC) cells
  • Used in epilepsy
  • Used in bipolar disorder
  • Used in metabolic disease
  • Used in HIV infection
  • Used for the prevention of migraine headaches