Learn More
Medchemexpress LLC Valproic acid-d14 so 1mg | 1MG
SDP
Catalog No. 5000243143
Encompass_Preferred
Valproic acid-d14 (sodium) is deuterium labeled Valproic acid (sodium) Valproic acid sodium salt (Sodium Valproate) is an HDAC inhibitor with IC50 in the range of 0 5 and 2 mM also inhibits HDAC1 (IC50 400 M) and induces proteasomal degradation of HDAC2 Valproic acid sodium salt activates Notch1 signaling and inhibits proliferation in small cell lung cancer (SCLC) cells Valproic acid sodium salt is used in the treatment of epilepsy bipolar disorder and prevention of migraine headaches
Catalog No. 50-002-43143 Supplier Medchemexpress LLC Supplier No. HY10585AS11MG
Valproic acid-d14 (sodium) is deuterium labeled Valproic acid (sodium) Valproic acid sodium salt (Sodium Valproate) is an HDAC inhibitor with IC50 in the range of 0 5 and 2 mM also inhibits HDAC1 (IC50 400 M) and induces proteasomal degradation of HDAC2 Valproic acid sodium salt activates Notch1 signaling and inhibits proliferation in small cell lung cancer (SCLC) cells Valproic acid sodium salt is used in the treatment of epilepsy bipolar disorder and prevention of migraine headaches